Anti-EIF4G1 Goat Polyclonal Antibody

Supplier: R & D Systems
AF4018
220011-424EA 663.78 USD
220011-424
Anti-EIF4G1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EIF4G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR