Anti-FGF21 Goat Polyclonal Antibody

Supplier: R & D Systems
AF2539
220010-584EA 707.17 USD
220010-584
Anti-FGF21 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FGF21
Antigen Name: fibroblast growth factor 21
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR