Anti-C5 Goat Polyclonal Antibody

Supplier: R & D Systems
AF2037
220010-261EA 692.71 USD
220010-261
Anti-C5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR