Anti-TNFRSF17 Goat Polyclonal Antibody

Supplier: R & D Systems
AF193
220010-199EA 707.17 USD
220010-199
Anti-TNFRSF17 Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR