Anti-TMPRSS15 Goat Polyclonal Antibody

Supplier: R & D Systems
AF1585
220009-990EA 663.78 USD
220009-990
Anti-TMPRSS15 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TMPRSS15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR