Anti-IL17C Goat Polyclonal Antibody

Supplier: R & D Systems
AF1234
220009-791EA 663.78 USD
220009-791
Anti-IL17C Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL17C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR