Anti-GABRB2 Goat Polyclonal Antibody

Supplier: R & D Systems
AF1188
220009-762EA 649.32 USD
220009-762
Anti-GABRB2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GABA-B R2
Antigen Name:
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR