Surgi-OR™ Russian Forceps, Mid-Grade, Sklar
Supplier: Sklar
95-604 95-607
10184-618EA 46.97 USD
10184-618 10184-622
Surgi-OR™ Russian Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Russian Forceps are medium-heavy forceps used for grasping heavy or thick tissue
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
They are also used during wound closure.