Bone Anchor Screws, Stoelting
Supplier: Stoelting
51462 51457 51458 51456 51459
95060-198PK 90.04 USD
95060-198 100229-628 100229-626 MSPP-51462 100229-622
Bone Anchor Screws, Stoelting
Fasteners Screws
Bone Anchor Screws anchor cannulae or other chronically-attached appliances to skull.
  • Ideal for use in stereotaxic surgeries
  • One pound of dental cement and 8 oz. of solvent
Dental cement is used to affix stainless steel to skull or bone, for lab repairs or for casting of hard acrylic parts and are particularly useful for securing bone anchor screws in skull holes or cementing cannulae in place. Dental cement can be cut, drilled or ground and the mixture is exothermic.