Boreal Science Stereomicroscope Bundles
Supplier: Avantor
470218-188KT 1731.69 USD
470218-188 470218-190
Boreal Science Stereomicroscope Bundles
Microscopes
Special Offer! Buy 5 Get 1 Free!
This Boreal Stereomicroscope, #5540400, features an inclined head for viewing comfort, full size of 30cm optics and an easy adjust collar for precise control of focus tension. With rack and pinion focusing, rugged die-cast metal construction, and a reagent resistant finish, this stereomicroscope is very durable. It has turret mounted 2x and 4x objectives for quick magnification changes, and a dual illumination system for ideal specimen lighting. All Boreal Microscopes are backed by our All-inclusive Warranty that covers parts, labor and shipping costs with no restriction on ownership.
Don’t need 6? Buy 1 and get a Free Fossil Set!
Take advantage of this limited time offer. Buy 1 Boreal Stereomicroscope, #5540400, and receive our Introductory Fossil Collection, #952100 Free! This collection is a fine introduction to the classification of fossil animals and plants. Fossils from nine different phyla form a wide range of geological periods and are all carefully selected to illustrate important life forms and different modes of preservation.
  • Special bundle offers
  • Buy 5, get 1 free
  • Buy 1 and get a free fossil set
  • Boreal lifetime warranty
This Boreal Science Stereomicroscope, model 5540400, features an inclined head for viewing comfort, full size 30cm optics and an easy adjust collar for precise control of focus tension. With rack and pinion focusing, rugged diecast metal construction and a reagent finish, this stereomicroscope is very durable. It has turret mounted 2x and 4x objectives for quick magnification changes and the dual illumination is ideal for specimen lighting. All Boreal microscopes are backed by our All-inclusive Warranty that covers parts, labor and shipping costs with no restriction on ownership.
Don’t need 6? Buy 1 and get a Free Fossil Set!
Take advantage of this exceptional offer. Buy 1 Boreal Science Stereomicroscope, model 5540400, and receive our Introductory Fossil Collection, model 500110 Free! This collection is a fine introduction to the classification of fossil animals and plants. Fossils from nine different phyla form a wide range of geological periods and are all carefully selected to illustrate important life forms and different modes of preservation.