tert-Butyl hypochlorite ≥98%

Supplier: Pfaltz & Bauer
Danger

Synonyms: Hypochlorous Acid tert-Butyl Ester

B29310-25G
101186-480EA 1658.15 USD
101186-480
tert-Butyl hypochlorite ≥98%
tert-Butyl hypochlorite
Formula: C₄H₉ClO
MW: 108.57 g/mol
Boiling Pt: 78 °C
Density: 0,96
MDL Number: MFCD00037105
CAS Number: 507-40-4
EINECS: 208-072-7
UN: 3255
ADR: 4.2, 8,I

Order Now

Specification Test Results

Content 98%
Flash Point −10°C
Storage Temperature 2 to 8°C
Sensitivities Moisture, Light, Air
Appearance Clear Yellow Liquid
Boiling Point 78°C
Density 0.96 g/cm3
Refractive Index 1.398
Stability Stable under normal temperatures and pressures

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR