Copper citrate hemipentahydrate ≥98%

Supplier: Pfaltz & Bauer
Warning

C25320-100G
101180-486EA 1496.03 USD
101180-486
Copper citrate hemipentahydrate ≥98%
Copper citrate hemipentahydrate
Formula: C₆H₄Cu₂O₇·⁵/₂H₂O
MW: 360.00 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00150504
CAS Number: 10402-15-0
EINECS: 212-752-9

Order Now

Specification Test Results

Synonyms Dicopper 2-oxidopropane-1,2,3-tricarboxylate hemipentahydrate
Content 98%
Storage Temperature 15 to 30°C
Appearance Green to Blue Crystalline Powder
Odor Odorless
Melting Point 300°C
Solubility in Water Slightly Soluble

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR