Surgi-OR™ Mayo-Hegar Needle Holder, Mid Grade, Sklar
Supplier: Sklar
95-842 95-847 95-853 95-869
89023-732EA 40.02 USD
89023-732 89023-734 89023-736 89023-738
Surgi-OR™ Mayo-Hegar Needle Holder, Mid Grade, Sklar
Forceps
The Surgi-OR™ Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures
Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is used with larger, heavier suture needles because of it's wide jaws.
Surgi-OR™ instruments are mid-grade instruments, suitable for use in the operating room or surgery center.
Due to its versatility, it is one of the most commonly used surgical tools for both human and animal procedures.