Nail Splitters, OR Grade, Sklar
Supplier: Sklar
97-1660 97-1661 97-1663 97-1662
10184-798EA 294.58 USD
10184-798 10194-166 10194-036 10194-038
Nail Splitters, OR Grade, Sklar
Cutters
Nail Splitter is a podiatric instrument used for making sectional, longitudinal cuts in nails as a part of nail surgery.
  • It is a double-spring, plier handle instrument with tapered jaws.