Surgi-OR™ TC Mayo-Hegar Needle Holder, Mid Grade, Sklar®
Supplier: Sklar
95-140 95-141 95-142 95-143
89023-504EA 150.28 USD
89023-504 89023-506 89023-508 89023-510
Surgi-OR™ TC Mayo-Hegar Needle Holder, Mid Grade, Sklar®
Forceps
The Surgi-OR™ TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures
Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision.
Surgi-OR™ instruments are mid-grade instruments, suitable for use in the operating room or surgery center.
Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procedures.