Self-Contained Biological Indicator Ampoules, Mesa Laboratories

Supplier: Mesa Labs
77-02-PS-6-50 77-02-PS-4-10 77-02-PS-4-50 77-02-PS-5-10 77-02-PS-5-50
101384-680PK 501.51 USD
101384-680 101384-672 101384-674 101384-676 101384-678
Self-Contained Biological Indicator Ampoules, Mesa Laboratories
Sterilization Indicators
ProSpore Ampoule are self-contained biological indicators for steam sterilization ideal for validating the sterilization of liquid loads, though they are also suitable for loads containing dry goods. Simply expose the unit to the sterilization process and incubate afterwards. They are validated for 48 hour reduced incubation.

  • No activation is required
  • Hermetically sealed, type I borosilicate ampoule
  • Negative controls available (for 1 ml)
  • Use with industrial steam sterilization processes between 118 – 135 °C

The negative control is a media ampoule made from the same lot of media used to make the corresponding spore ampoules. The ampoule is filled with a modified soybean casein digest broth containing bromocresol purple acid indicator. Each ampoule also contains a population of G. stearothermophilus spores. Growth is evident by either turbidity and/or a color change from purple to or toward yellow.

Ampoule outer diameter: 14.75 ± 0.15 mm

Ampoule height: 52 – 55 mm

Certifications: Complies to AAMI/ISO 11138-1 and AAMI/ISO 11138-3.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR