Sklarlite™ Webster Needle Holder, OR Grade, Sklar
Supplier: Sklar
23-2120 23-2125
10195-318EA 269.86 USD
10195-318 10195-322
Sklarlite™ Webster Needle Holder, OR Grade, Sklar
Forceps
Sklarlite™ Webster Needle Holders are used with small needles and fine suture material, in plastic surgery or subcutaneous procedures.
Smooth jaws will cause the least damage to the needle, however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments, while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.