Bromine ≥99.5%, Laboratory Reagent, VWR Chemicals BDH®

Supplier: BDH
Danger

S051201-VWNK03 S051201-VWNF03
BDH4576-500MLEA 866.59 USD
BDH4576-500ML BDH4576500ML
Bromine ≥99.5%, Laboratory Reagent, VWR Chemicals BDH®
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7 °C (1013 hPa)
Melting Pt: –7.2 °C
Density: 3.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010896
CAS Number: 7726-95-6
EINECS: 231-778-1
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Specification Test Results

Assay Min. 99.5 %
Heavy metals (as Pb) Max. 2 ppm
Residue after evaporation Max. 50 ppm
Cl (Chlorine) Max. 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR