Water for LC-MS, Pierce™
Supplier: Thermo Fisher Scientific
Synonyms:
Dihydrogen oxide
Pierce
T51140-K2
51140
PI51140EA
96.63
USD
CAPI51140
PI51140
Water for LC-MS, Pierce™
Water
Thermo Scientific Pierce Water, LC-MS Grade is an ultrapure, LC-MS grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.
- Purity and quality validated with 30 tests for use in LC-MS applications
- 0.2 micron filtered, packed in solvent-rinsed amber glass bottles with Teflon TFE-lined fluorocarbon caps for ultimate protection
- Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases
- High lot-to-lot consistency and reliability
Low UV absorptivity makes this product ideal for use in HPLC applications.
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...