Pierce™ Cypridina Luciferase Glow Assay Kit, Thermo Scientific
Supplier: 0000013509
16170 16171
PI16170EA 158.4 USD
PI16170 PI16171
Pierce™ Cypridina Luciferase Glow Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Cypridina Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life greater than one hour) in the presence of Cypridina luciferase reporters.
  • Sensitive—highly sensitive detection of Cypridina luciferase activity
  • Secreted luciferase—allows real-time assays and kinetic studies without destroying cells
  • Compatible—assay reagents compatible with other Cypridina or Vargula luciferases
  • Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
This Pierce Luciferase Glow Assay Kit contains reagents for measuring Cypridina luciferase activity in mammalian cell culture media and lysates. The light output generated by the luciferase reaction can be correlated to the amount of luciferase protein produced, which in turn is proportional to the activity of the promoter driving the luciferase expression.
Delivery information: Kit contains Cypridina glow assay buffer (5 or 50 ml); 1X vargulin (5 µl); and 2X cell lysis buffer (6 or 60 ml).
Caution: For Research Use Only. Not for use in diagnostic procedures.