Pierce™ Antibody Coated Plates, 96 Well,, Thermo Scientific
Supplier: Thermo Scientific
Pierce
15137 15136 15134 15234 15134 15137 15234 15135 15135
PI15137EA 216 USD
CAPI15134 PI15137 PI15136 PI15234 PI15135 PI15134 CAPI15234 CAPI15135 CAPI15137
Pierce™ Antibody Coated Plates, 96 Well,, Thermo Scientific
Microplates Immunology Microplates and Strips
Pierce™ Antibody coated plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
  • Antibody-binding capacity is higher than direct adsorption onto polystyrene
  • Ideal for direct, indirect, competitive or sandwich assays
  • Activation level: 100 µL/well
  • Pre-blocked with SuperBlock™ buffer to reduce non specific binding
Clear plates for colorimetric ELISA and other assays. White plates for chemiluminescent. Black plates for fluorescent assays.
Caution: For Research Use Only. Not for use in diagnostic procedures.