NCP Series Nitrile Co-Polymer Powder Free Class 100 Gloves, CT International
Supplier: CT International
NCP201 NCP202
56613-858CS 408.36 USD
56613-858 56613-860
NCP Series Nitrile Co-Polymer Powder Free Class 100 Gloves, CT International
Gloves
NCP Series gloves are class 100 gloves are manufactured for outstanding strength, stretch and fit.
  • Offer a textured surface and a super soft feel for comfort
  • Nitrile co-polymer gloves
  • Powder free
Certifications: These gloves are ISO 5 clean room compatible or higher controlled environments.