Sodium metabisulfite ≥97.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
di-Sodium disulfite
, Disodiumdisulphite
, Disodium disulfite
, Sodium disulphite
, Disodium disulphite
, Sodium pyrosulphite
, Sodium disulfite
, Disodiumdisulfite
, di-Sodium disulphite
, Sodium pyrosulfite
BDHVBDH9282-500G
BDH9316-500GCS
685.81
USD
BDH9316-500G
Sodium metabisulfite ≥97.0% ACS, VWR Chemicals BDH®
Sodium metabisulphite
Formula:
Na₂S₂O₅ MW: 190.11 g/mol Melting Pt: 150 °C Density: 1.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 EINECS: 231-673-0 REACH: 01-2119531326-45 Merck Index: 12,08784 |
Specification Test Results
Chloride |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Assay |
|
Thiosulfate |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...