Tartrazine 1.5% saturated solution in Cellosolve

Supplier: Electron Microscopy Sciences

Synonyms: FD and C Yellow No. 5Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylateAcid Yellow 23Sodium 5-oxo-1-(4-sulfonatophenyl)-4-((4-sulfonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylateFilter Yellow

21742
101410-826EA 132.43 USD
101410-826
Tartrazine 1.5% saturated solution in Cellosolve
Tartrazine
Formula: C₁₆H₁₂N₄O₉S₂
MW: 534.36 g/mol
MDL Number: MFCD00148908
CAS Number: 1934-21-0
https://us.vwr.com/stibo/web/std.lang.all/49/13/28984913.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR