Acetic acid glacial ≥99.7% ACS
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
10040
10041
102090-954EA
109.4
USD
102090-954
102090-956
Acetic acid glacial ≥99.7% ACS
Acetic acid
Reagent A.C.S Acetic acid. C2H4O2 F.W. 60.05, with an assay minimum of 99.7%.
Formula:
H₃CCOOH Boiling Pt: 118 °C (244 °F) Melting Pt: 16.6 °C (62 °F) Density: 1.05 g/cm³ (8.762 lbs/gal.) Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8,II |
|
Specification Test Results
Assay | 99.7% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...