Jenner's stain solution stain for hematology

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Eosin−Methylene Blue

26033-05
101410-898EA 89.49 USD
101410-898
Jenner's stain solution stain for hematology
Jenner's stain

Jenner Stain solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology.


  • High quality staining solution
  • Ready-to-use

Flash Pt: 11 °C (51.8 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081733
CAS Number: 62851-42-7
EINECS: 200-659-6
UN: 1230
ADR: 3,6.1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR