Masson's Trichrome for Connective Tissue, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26367-01
26367-05
26367-03
26367-04
26367-06
26367-07
26367-02
101411-278EA
53.46
USD
101411-278
101411-286
101411-282
101411-284
101411-288
101411-290
101411-280
Masson's Trichrome for Connective Tissue, Electron Microscopy Sciences
Stains
Histology Stains
Masson’s Trichrome for Connective Tissue, Sheehan and Hrapchak (1980), for staining nuclei, agentaffen granules, collagen, cytoplasm, keratin, etc
Stain results - Nuclei: Black, Cytoplasm, Keratin, Muscle Fibers: Red, Collagen, Mucin: Blue.