Acrylic Plastic Casting, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
24210-01 24210-02 24210-08
101381-610EA 81.32 USD
101381-610 101381-612 101381-614
Acrylic Plastic Casting, Electron Microscopy Science
Specimen Mounting Supplies
Acrylic plastic casting is a thermosetting acrylic-polymer plastic with excellent clarity
Acrylic Plastic Casting: Thermosetting acrylic-polymer plastic with excellent clarity. Place object in a mold, mix and pour. Hard in an hour and object is firmly embedded in clear, stable plastic. Can be polished with soft cloth to high gloss.
Delivery information: The kit consisits of resin and hardener.