Modified Movat’s Stain, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26385-10
26385-03
26385-02
26385-05
26385-04
26385-09
26385-07
26385-06
26385-08
26385-01
100504-624EA
89.13
USD
100504-624
100504-610
100504-608
100504-614
100504-612
100504-622
100504-618
100504-616
100504-620
100504-606
Modified Movat’s Stain, Electron Microscopy Sciences
Stains
Histology Stains
Modified Movat’s Stain, is used for connective tissue stains – Elastic tissue stains
Stain results - Nuclei: Black, Cytoplasm: Red, Elastic Fibers: Purple to Black, Collagen and Bone: Yellow, Mucopolysaccharides: Blue-Green, Muscle: Red.