Wright's stain solution stain for hematology

Supplier: Electron Microscopy Sciences

26060-05 26060-06
100504-442EA 77.48 USD
100504-442 100504-444
Wright's stain solution stain for hematology
Wright's stain

For use as a blood stain


  • High quality staining solution
  • Ready-to-use


For use as a blood stain. Ready-to-use.

MDL Number: MFCD00082143
CAS Number: 68988-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7542c58f3a614807fa06d88237237b0aa9b88b22&vid=aa60fa7264f7d21e0ef4f98e1d904c8b4fb44f5a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR