Water ACS, deionized
Supplier: Electron Microscopy Sciences
22800-05
22800-01
100504-380EA
118.35
USD
100504-380
100504-378
Water ACS, deionized
Water
EMS reagent grade water is typically prepared at 18 megohm/cm specific resistance using a reverse osmosis, mixed deionization, activated filtration and final filtration at 0.2 microns.
Formula:
H₂O Boiling Pt: 100 °C (212 °F) Melting Pt: 0 °C (32 °F) Density: 1 g/cm³ (8.345 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Specification Test Results
Bacteriological purity | 0 CFUs/l |
Residue after evaporation | 10 ppm |
Coliform | negative |
Color (APHA) | <+/-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...