Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) free base ≥99.9%, Reagent Grade

Supplier: Electron Microscopy Sciences
Warning

11720 11700
100503-720EA 132.02 USD
100503-720 100504-922
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) free base ≥99.9%, Reagent Grade
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Tris (Hydroxymethyl) Aminomethane Free Base, Reagent Grade is a white crystalline powder. A 40% (w/v) aqueous solution. Hydroxymethyl Aminomethane. C4H11NO3 F.W. 121.14 CAS 77-86-1 A 40% (w/v) aqueous solution. It is clear and colorless. Assay 99.9% Water .0.2% Heavy Metals.-2ppm


Solubility: 40% (w/v) aqueous solution is clear and colorless

Formula: NH₂C(CH₂OH)₃
Density: 0.84159 g/cm³ (7.023 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >99.9%
Heavy metals 2 ppm
Water <0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR