Succinic acid ≥99.0%, crystals ACS
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
1,4-Butanedioic acid
21550
100504-302EA
50.2
USD
100504-302
Succinic acid ≥99.0%, crystals ACS
Succinic acid
Formula:
HOOCCH₂CH₂COOH MW: 118.09 g/mol Boiling Pt: 235 °C (455 °F) Melting Pt: 184 °C (363 °F) Density: 1.564 g/cm³ (13.052 lbs/gal.) Storage Temperature: Ambient |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 UN: 3261 ADR: 8,III |
Specification Test Results
Assay |
|
Chloride | 0.001% |
Heavy metals | 5 ppm |
Insoluble matters | 0.01% |
Iron | 5 ppm |
Nitrogen compounds | 0.001% |
Phosphate | 0.001% |
Sulfate | 0.003% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...