Orange G, certified (C.I. 16230)

Supplier: Electron Microscopy Sciences

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonateAcid Orange 10Wool Orange 2GOrange G sodium saltCI 16230

19070
100504-126EA 62.48 USD
100504-126
Orange G, certified (C.I. 16230)
Orange G

Kornhauser's quad stain for most elementary structures of tissues. Stain Tech., 18, 985 (1943) Mallory's tissue stain for connective tissue. Mallory, J. Exper. Med., 5,15 (1900). Used with safranin and tannic acid for showing shoot apex in plant tissue. Stain Tech., 18, 105 (1943).

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
https://us.vwr.com/stibo/web/std.lang.all/48/69/28984869.jpg

Order Now

Specification Test Results

Dye Content Approx. 60%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR