Ninhydrin, crystals, Reagent Grade

Supplier: Electron Microscopy Sciences
Warning

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

19030
100504-122EA 131.88 USD
100504-122
Ninhydrin, crystals, Reagent Grade
Ninhydrin

(1,2,3-Triketohydrindene Monohydrate)
C9O4·H2O F.W. 178.15 CAS 485-47-2

Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 449.3 °C (841 °F)
Melting Pt: 250 °C (482 °F)
Density: 1.714 g/cm³ (14.303 lbs/gal.)
Flash Pt: 240 °C (464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
https://us.vwr.com/stibo/web/std.lang.all/48/65/28984865.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR