May Grunwald's eosin-methylene blue solution in methanol
Supplier: Electron Microscopy Sciences
|
Danger
|
26064-06
26064-05
100504-446EA
75.85
USD
100504-446
100504-448
May Grunwald's eosin-methylene blue solution in methanol
May Grunwald's eosin-methylene blue
A ready-to-use stain for bone marrow staining.
- High quality staining solution
- Ready-to-use
Boiling Pt:
64.7 °C (148 °F) Melting Pt: –98 °C (–144 °F) Density: 0.79 g/cm³ (6.593 lbs/gal.) Flash Pt: 11 °C (52 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00131580 UN: 1230 ADR: 3,II |