Light green SF yellowish solution_SC 0,2% acetic acid_0,2

Supplier: Electron Microscopy Sciences

Synonyms: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Dihydrogen (ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulfonatobenzyl)ammonium disodium salt, Light Green 2G, S or 2GN, CI 42095, acid Green 5, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonate, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, acid Green F, CN DcL-30

26085-05 26085-06
101410-980EA 62.46 USD
101410-980 101410-982
Light green SF yellowish solution_SC 0,2% acetic acid_0,2
Light Green SF Yellowish

Light Green SF Yellowish is a high quality ready to use solution. The solution is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a fungal stain and in a 1% aqueous solution.

Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
MDL Number: MFCD00012121
CAS Number: 5141-20-8
Merck Index: 13,05506
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53e83ad8dde11e21d14f5efdff8d7d0d23b9f02e&vid=c489b6d7ffbba967f532d0757c7fc2c4f87f497b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR