Light green SF yellowish, certified (C.I. 42095, C.N. DcL-30)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: CI 42095Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonateDihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium saltSodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonateacid Green FLight Green 2G, S or 2GNDihydrogen (ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulfonatobenzyl)ammonium disodium saltacid Green 5Fast acid Green NCN DcL-30Food Green n°2

17920
100504-078EA 30.62 USD
100504-078
Light green SF yellowish, certified (C.I. 42095, C.N. DcL-30)
Light Green SF Yellowish

Grocott's method for fungi. Dahl's method as a bone and calcium stain. McManus' method for glycogen. Fraser-Lendrum method as a connective tissue stain.


Solubility : 20.35% Water; 4.0% Alcohol; 5.25% Cellosolve; 12.0% Glycol; 0.0% Xylene


Certifications: Certified by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
MDL Number: MFCD00012121
CAS Number: 5141-20-8
https://us.vwr.com/stibo/web/std.lang.all/48/43/28984843.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR