Indigo carmine, certified (C.I. 73015)

Supplier: Electron Microscopy Sciences
Warning

Synonyms: CI 730155,5'-Indigodisulfonic acid, Disodium saltCarmine BlueFood blue No. 2Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonateSodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulfonateacid Blue 745,5'-Indigodisulphonic acid, Disodium saltFD and C Blue No. 2FD&C Blue #2

16910
100504-058EA 102.73 USD
100504-058
Indigo carmine, certified (C.I. 73015)
Indigo carmine

Shunway's stain for animal embryos. Used with Acid Fuchsin for staining Negri bodies (Stain Tech., 5, 34 (1928)) and counterstain to hematoxylin for staining vaginal smears (Am. J. Anat., 61, 505 (1937)).

Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Density: 450 kg/m³
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
https://us.vwr.com/stibo/web/std.lang.all/48/40/28984840.jpg

Order Now

Specification Test Results

Dye content Approx. 85%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR