Formic acid ≥96% ACS

Supplier: Electron Microscopy Sciences

Synonyms: Aminic Acid, Methanoic acid

15760 15750
100503-932EA 133.08 USD
100503-932 100503-930
Formic acid ≥96% ACS
Formic acid

HCOOH F.W. 46.03x Specific Gravity: 1.22

Formula: HCO₂H
MW: 46.03
Boiling Pt: 101 °C (214 °F)
Melting Pt: –9 °C (16 °F)
Density: 1.6 g/cm³ (13.352 lbs/gal.)
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
UN: 3412
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609771e5ed4eb14b6e4ed45a476b18d2f75e880e&vid=609771e5ed4eb14b6e4ed45a476b18d2f75e880e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity 1.22


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR