2-Ethoxyethanol ≥99.5%, purified

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve

15310-01 15310-06
100496-482EA 85.34 USD
100496-482 100496-484
2-Ethoxyethanol ≥99.5%, purified
2-Ethoxyethanol

Suitable for histological use.

Formula: C₂H₅OCH₂CH₂OH
MW: 90.12 g/mol
Boiling Pt: 135
Melting Pt: -70
Density: 0.93 g/cm³ (7.761 lbs/gal.)
Flash Pt: 40 °C (104 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002869
CAS Number: 110-80-5
UN: 1171
ADR: 3 III
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90ea1e005a7c622b792d1e14526d3542a9c83a6d&vid=5bbe9d57a41c250e9d935a55a0c081db1dfb6921&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay >99.5%
Residue After Evaporation <0.001%
Specific Gravity at 20 °C 0.931
Water <0.1%
Boiling Point 133-135°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR