Ethanol denatured ≥95% (denatured with 5% methanol)

Supplier: Electron Microscopy Sciences
Danger

15051 15050
100505-052EA 174.56 USD
100505-052 100505-050
Ethanol denatured ≥95% (denatured with 5% methanol)
Ethanol denatured

Blended 95% ethanol and 5% methanol, Suitable for all routine histology/cytology preparations.

Formula: H₃CCH₂OH
Boiling Pt: 79 °C (174 °F)
Density: 0.7905 g/cm³ (6.597 lbs/gal.)
Flash Pt: 14 °C (57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Evaporation Residue 5 ppm max.
Filtered SDA-3A 95%
Water 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR