Erythrosin B disodium salt, certified (C.I. 45430)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: FD&C Red #3Erythrosin 239Sodium 2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Erythrosine 3BErythrosine sodiumC.I. 45430Erythrosin BAcid Red 51

15010
100503-888EA 55.15 USD
100503-888
Erythrosin B disodium salt, certified (C.I. 45430)
Erythrosin B disodium salt

Used with methylene blue as a plasma stain for nerve cells and use crystal violet for plant tissue.


Solubility : 10.0% Water; 5.0% Alcohol; 7.0% Cellosolve; 8.25% Glycol; 0.0% Xylene

Formula: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Density: 1 g/cm³ (8.345 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
UN: 2810
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/48/16/28984816.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR