Chloroform ≥99.7%, Glass Distilled

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

12551 12550
100503-762EA 96.67 USD
100503-762 100503-760
Chloroform ≥99.7%, Glass Distilled
Chloroform

Glass distilled chloroform is available for your convenience.

Formula: CHCl₃
Boiling Pt: 62 °C (144 °F)
Melting Pt: –63 °C (–81 °F)
Density: 1.47988 g/cm³ (12.35 lbs/gal.)
Flash Pt: 0 °C (32 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd7b089b86eb10623b4c77ccca3213cbc0d47c83&vid=f01efdcdf34b24309a77803e09d1562bc60047ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7% min
Evaporation Residue 3 ppm max.
Water 0.03% max.
pH range 4.6 - 6.2
UV Absorption 1 cm cell vs H₂O 290 nm: 0.01, 270 nm: 0.02, 260 nm: 0.05, 255 nm: 0.15, 245 nm: 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR