Brilliant cresyl blue, certified (C.I.51010)

Supplier: Electron Microscopy Sciences
Danger

Synonyms: BCBBrilliant Blue CBrilliant cresyl blue zinc chloride double salt1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1)CI 51010

11465
100504-920EA 103.66 USD
100504-920
Brilliant cresyl blue, certified (C.I.51010)
Brilliant Cresyl Blue

Brilliant Cresyl Blue, Certified C.I. DcV-5 is also known as Brilliant Blue C and Cresyl Blue 2RN.


Major uses: Platelets and reticulum of immature red cells. Counterstained with Wright's stain.


Solubility : 3.0% Water; 2.6% Alcohol; 2.25% Cellosolve; 8.0% Glycol; 9.5% Xylene


Certifications: Certified C.I. DcV-5 by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
MDL Number: MFCD00148901
CAS Number: 10127-36-3
UN: 3288
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/47/91/28984791.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR