Bismarck brown Y, certified C.I. 21000

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Phenylene BrownBasic Brown 14,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochlorideVesuvineLeather BrownC.I. 21000Manchester BrownBasic Brown G, GX, GXPBismarck Brown Y (G)1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride

11310
100503-688EA 56.81 USD
100503-688
Bismarck brown Y, certified C.I. 21000
Bismarck Brown Y

PAP for staining smears. Nuclei and granules. Mucin and calciform cells of intestine, cartilage and embryo.
Platelets & reticulum of immature red cells.


Can be used as counterstained with Wright's stain.


Solubility : 1.5% Water; 3.0% Alcohol; 3.0% Cellosolve; 7.9% Glycol; 0.05% Xylene


Certifications: Certified by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₁₈H₁₈N8·2HCl
MW: 419.32 g/mol
MDL Number: MFCD00012977
CAS Number: 10114-58-6
https://us.vwr.com/stibo/web/std.lang.all/47/89/28984789.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR