Hydrochloric acid 1% alcohol_95
Supplier: Electron Microscopy Sciences
Synonyms:
Hydrochloric acid solution, Muriatic acid
26073-04
26073-01
100504-460EA
275.05
USD
100504-460
100504-458
Hydrochloric acid 1% alcohol_95
Hydrochloric acid
An acidified alcohol solution for the differentiation of regressive stains.
Formula:
HCl MW: 36.46 |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...