1,2-Dichloroethane ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Ethylene chloride, Ethylene dichloride, Dichloroethane

13251 13250
100503-816EA 229.11 USD
100503-816 100503-814
1,2-Dichloroethane ACS
1,2-Dichloroethane

(Ethylene Dichloride) CH2ClCH2Cl F.W. 98.96 CAS 107-06-2 A.C.S. Specifications

Formula: ClH₂CCH₂Cl
MW: 98.96 g/mol
Boiling Pt: 83.5
Melting Pt: -35.5
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
UN: 1184
ADR: 3 II
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2de42d5d4158bf6cabf4ec0d4bbb58630389f7d0&vid=2de42d5d4158bf6cabf4ec0d4bbb58630389f7d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Boiling Range, Incl. 83.5 °C 20 °C
Odor Resembling odor of CHCl₃
Residue after evaporation 0.002%
Titratable acid 0.0003 meq/g
Water 0.03%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR