2-(Dimethylamino)-ethanol

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 2-DimethylaminoethanolN,N-DimethylethanolamineDMAE

13300
100503-826EA 11.56 USD
100503-826
2-(Dimethylamino)-ethanol
2-Dimethylaminoethanol

A curing agent for epoxy resins.

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 133 °C (271.4 °F)
Melting Pt: −70… −94 °CF
Density: 0.89 g/cm³ (7.42705 lbs/gal.)
Flash Pt: 31 °C (87.8 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
UN: 2051
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/48/12/28984812.jpg

Order Now

Specification Test Results

Specific Gravity 0.883 - 0.888

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR