Merit™ Hartmann Mosquito Forceps, Physician Grade, Sklar
Supplier: Sklar
97-409 97-421
10184-434EA 21.34 USD
10184-434 10192-290
Merit™ Hartmann Mosquito Forceps, Physician Grade, Sklar
Forceps
Merit™ Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Merit™ instruments are office-grade line of products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
Used in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures.